Write Protect Tape, Electron Microscopy Sciences
Product Details & Documents
Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers.
- Will not dry out, peel or fall off
- Withstand temperatures ranging from –196 to 150 °C
The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 and 1.5 mL tubes.
Specifications
- Description:Write Protect Tape
- Dimensions:32 x 13 mm