Orders LinkContactUsLinkComponent
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Catalog # 102098-702
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Catalog # 102098-702
Supplier:  
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Labels for moist, frosty or frozen tubes.
  • Temperature resistant to –40 °C
Direct thermal Labels are white and on a 1" int.Ø. core. Temperature Range: –40 to +70 °C.
GRIPS to wet, frost-covered and frozen surface, Compatible with centrifuge rotors, Resistant to chemicals and solvents, Adheres to plastics, glass, metals and even paperboard boxes.
Print on sheets with Laser printers or use rolls for direct thermal printers.