Orders LinkContactUsLinkComponent
JEOL SEM Mount, Electron Microscopy Sciences
JEOL, 32 x 5 mm, Aluminum
Catalog # 102098-252
 :  75830-60
JEOL SEM Mount, Electron Microscopy Sciences
Catalog # 102098-252
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
JEOL, 32 x 5 mm, Aluminum
Price
$380.53
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications
  • Description:
    JEOL
  • Material:
    Aluminum
  • Size:
    32 x 5 mm