Orders LinkContactUsLinkComponent
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 32 mm (1¼"), 10 mm (⅜")
  102098-210
 :  75635-95
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
  102098-210
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 32 mm (1¼"), 10 mm (⅜")
$511.17
Pack of 100
  • Description:
    Hitachi M4
  • Height:
    10 mm (⅜")
  • Material:
    Aluminum
  • Head diameter:
    32 mm (1¼")