Orders LinkContactUsLinkComponent
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 12 divisions, Aluminum, 32 mm (1¼"), 6.0 mm (¼")
Catalog # 102098-208
 :  75635-90D
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Catalog # 102098-208
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 12 divisions, Aluminum, 32 mm (1¼"), 6.0 mm (¼")
Price
$1,352.74
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications
  • Description:
    Hitachi M4 12 divisions
  • Height:
    6.0 mm (¼")
  • Material:
    Aluminum
  • Head diameter:
    32 mm (1¼")