Orders LinkContactUsLinkComponent
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 50 mm (2"), 6.0 mm (¼")
Catalog # 102098-204
 :  75635-87
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Catalog # 102098-204
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 50 mm (2"), 6.0 mm (¼")
Price
$444.29
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications
  • Description:
    Hitachi M4
  • Height:
    6.0 mm (¼")
  • Material:
    Aluminum
  • Head diameter:
    50 mm (2")