Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Product Details & Documents
Specifications
- Description:Hitachi M4 12 divisions
- Height:6.0 mm (¼")
- Material:Aluminum
- Head diameter:32 mm (1¼")