Orders LinkContactUsLinkComponent
Digital Vortex-Genie 2, Electron Microscopy Sciences
Digital Vortex Genie 2, 120 V, 50 Hz Australian Plug
  102097-714
 :  71838-AU
Digital Vortex-Genie 2, Electron Microscopy Sciences
  102097-714
Digital Vortex-Genie 2, Electron Microscopy Sciences
Digital Vortex-Genie 2, Electron Microscopy Sciences
Digital Vortex Genie 2, 120 V, 50 Hz Australian Plug
$983.01
Each
Additional accessories for the Vortex Genie® 2 Digital available.
Features digital control and display. Uses shaking or vortexing action. Variable speed control allows slow speed shaking action up to high-speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. Multi-Task Versatility. A broad range of attachments of various shapes, sizes, and materials allows for mixing of almost all common tubes or containers for Hands-Free or Touch operation. Digital speed control and display. Provides accuracy, reproducibility and repeatability. Timer Function. Touch on for 1-99 seconds. Hands-Free for 1-99 minutes or continuous. Does not walk. Tough metal housing combined with "just enough weight", provides a stable platform for all types of mixing. Unmatched Reliability. Many years of proven performance in laboratories all over the world.
Additional accessories for the Vortex Genie® 2 Digital available.
  • Description:
    Digital Vortex Genie 2, 120 V, 50 Hz Australian Plug