Orders LinkContactUsLinkComponent
Slide Box-Foam, Electron Microscopy Sciences
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 100, Green
Catalog # 102097-568
Supplier #:  71478-14
Slide Box-Foam, Electron Microscopy Sciences
Catalog # 102097-568
Slide Box-Foam, Electron Microscopy Sciences
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 100, Green
Price
$16.82
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
  • Holds 25, 50, or 100 slides
  • Foam backing
  • Secure lock
  • Blue, green, red, yellow, white, black, and pink
Specifications
  • Capacity:
    100
  • Color:
    Green
  • Description:
    Slide Box