Orders LinkContactUsLinkComponent
Slide Box-Foam, Electron Microscopy Sciences
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 25, Red
  102097-546
Supplier #:  71478-03
Slide Box-Foam, Electron Microscopy Sciences
  102097-546
Slide Box-Foam, Electron Microscopy Sciences
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 25, Red
$9.50
Each
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
  • Holds 25, 50, or 100 slides
  • Foam backing
  • Secure lock
  • Blue, green, red, yellow, white, black, and pink
  • Capacity:
    25
  • Color:
    Red
  • Description:
    Slide Box