Orders LinkContactUsLinkComponent
Staining Dishes, Electron Microscopy Sciences
Steel lid only, 120×122×110 mm
  102097-522
 :  71424-L
Staining Dishes, Electron Microscopy Sciences
  102097-522
Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
Steel lid only, 120×122×110 mm
$76.11
Each
A variety of inexpensive staining dishes made from molded glass with and without fitted glass and stainless steel lids.
  • A variety of sizes are available
    Lids and dishes are available separately
Ideal for immersing slides in several solutions.
  • Description:
    Steel lid only
  • Dimensions:
    120×122×110 mm