Orders LinkContactUsLinkComponent
Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
Staining dish only, 120×122×110 mm
Catalog # 102097-518
Supplier #:  71424-D
Staining Dishes, Electron Microscopy Sciences
Catalog # 102097-518
Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
Staining dish only, 120×122×110 mm
Price
$138.55
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A variety of inexpensive staining dishes made from molded glass with and without fitted glass and stainless steel lids.
  • A variety of sizes are available
    Lids and dishes are available separately
Ideal for immersing slides in several solutions.
Specifications
  • Description:
    Staining dish only
  • Dimensions:
    120×122×110 mm