Cryogenic Grid Box, Electron Microscopy Sciences
Grid Box Round without Lid, Blue
Catalog # 102097-446
Supplier: Electron Microscopy Sciences
Supplier #: 71166-30
Cryogenic Grid Box, Electron Microscopy Sciences
Catalog # 102097-446
Supplier: Electron Microscopy Sciences
Jump to:
Product Details & Documents
These specimen grid boxes are for storing or transferring cryogenic TEM specimen grids.
- Four diamond shaped slots
- Non-static clear cover shield held in place with stainless steel screw, which is tapped in the center of the box
- Box only fits 71166-20 (Grid box square with lid)
- Available with lid, pin-type lid, or without lid
Documents
EMS-Core Sampling Tools
Product Literature
Specifications
- Color:Blue
- Description:Grid Box Round without Lid