Orders LinkContactUsLinkComponent
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
MEM-32 Grid Storage Box
  102097-440
Supplier #:  71165-10
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
  102097-440
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
MEM-32 Grid Storage Box
$218.83
Pack of 10
A newly designed Grid Box with safety, ease and convenience in mind. This multipurpose electron microscope specimen grid box is one of the most desirable boxes on the market.
Eliminates the chances of tweezer insertion damaging the grids – The "tweezers slot" and "grid slot" are in a separate location. The tweezers are only able to grip the edge of the grid enabling it to be picked up.
Grids stored no longer jump out of the box while you remove the cover – Between the body of the box and the sliding lid, there is a separate plastic cover, which allows for only four slots being exposed at a time.
The Grid Record Card is stored safely by insertion along on the reverse side of the box
MEM-96 – will able to store up to 96 grids
MEM-32 – will able to store up to 32 grids with 8 Blocks
Measures: 81 mm (L) x 54 mm (W) x 6 mm Thick
  • Description:
    MEM-32 Grid Storage Box