Orders LinkContactUsLinkComponent
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial A Grid-N-Block storage box with unique number
Catalog # 102097-430
Supplier #:  71158-20
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Catalog # 102097-430
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial A Grid-N-Block storage box with unique number
Price
$1,171.07
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Modules have 24 slots labeled with letters from A-X where the grids can be stored and rotated for easy access as well as 3 additional cavities for block storage .
Available with and without a unique identification number.
Specifications
  • Description:
    Dial A Grid-N-Block storage box with unique number