Orders LinkContactUsLinkComponent
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser Red
  102097-222
 :  70993-03
Parafilm Dispenser, Electron Microscopy Sciences
  102097-222
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser Red
$116.40
Each
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
  • Holds one or two 2" rolls, or one 4" roll
  • Size (W x H x D): 120 x 156 x 171 mm
  • Parafilm is a registered trademark of American can company
  • Description:
    Parafilm Dispenser Red