Orders LinkContactUsLinkComponent
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
Molded Flat Embedding Mold
  102097-198
Supplier #:  70905-12
Flat Embedding Molds, Electron Microscopy Sciences
  102097-198
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
Molded Flat Embedding Mold
$226.89
Pack of 12
Molds are injected molded polyethylene that is resistant to most embedding materials and will easily peeled-off the blocks. These molds are reusable.
  • Fits perfectly inside the cocoon box BE8
  • Each cavity measures 12L x 5W x 3.5D mm
  • Overall mold measures: 94 x 56 x 14 mm
  • Description:
    Molded Flat Embedding Mold