Orders LinkContactUsLinkComponent
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
Molded Flat Embedding Mold
Catalog # 102097-196
Supplier #:  70905-01
Flat Embedding Molds, Electron Microscopy Sciences
Catalog # 102097-196
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
Molded Flat Embedding Mold
Price
$21.64
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Molds are injected molded polyethylene that is resistant to most embedding materials and will easily peeled-off the blocks. These molds are reusable.
  • Fits perfectly inside the cocoon box BE8
  • Each cavity measures 12L x 5W x 3.5D mm
  • Overall mold measures: 94 x 56 x 14 mm
Specifications
  • Description:
    Molded Flat Embedding Mold