Flat Embedding with Convenient Box, Electron Microscopy Sciences
Product Details & Documents
No more stickiness with new storage box. Most resins are no longer a challenge to flat embed when used in combination with the Cocoon box. The system withstands temperatures up to 70 °C, and our flat embedding of many resins is made easier.
Specifications
- Description:3 mm Flat Embedding Mold with Cocoon Box