Orders LinkContactUsLinkComponent
Flat Embedding with Convenient Box, Electron Microscopy Sciences
Flat Embedding with Convenient Box, Electron Microscopy Sciences
4 mm Flat Embedding Mold with Cocoon Box
Catalog # 102097-190
Supplier #:  70900-CB
Flat Embedding with Convenient Box, Electron Microscopy Sciences
Catalog # 102097-190
Flat Embedding with Convenient Box, Electron Microscopy Sciences
Flat Embedding with Convenient Box, Electron Microscopy Sciences
4 mm Flat Embedding Mold with Cocoon Box
Price
$30.81
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
No more stickiness with new storage box. Most resins are no longer a challenge to flat embed when used in combination with the Cocoon box. The system withstands temperatures up to 70 °C, and our flat embedding of many resins is made easier.
Specifications
  • Description:
    4 mm Flat Embedding Mold with Cocoon Box