Orders LinkContactUsLinkComponent
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Capsules Size 00, 8 mm
Catalog # 102096-558
Supplier #:  70020-B
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Catalog # 102096-558
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Capsules Size 00, 8 mm
Price
$129.37
Log in for availability
Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These polyethylene embedding capsules have a 1 x 1 mm face at the tip of a truncated pyramid, each with a hinged cap.
Specifications
  • Ø:
    8 mm
  • Description:
    Embedding Capsules Size 00
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder, Clear
    Capsule Holder, Clear
    100490-714
    Log in for availability
    Pack of 6 - $146.65
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder, Clear
    Capsule Holder, Clear
    100490-716
    Log in for availability
    Pack of 12 - $287.65
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder for Size 00
    Capsule Holder for Size 00
    100490-718
    Log in for availability
    Pack of 6 - $147.26
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder for Size 00
    Capsule Holder for Size 00
    100490-720
    Log in for availability
    Pack of 12 - $238.04
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder, Clear
    Capsule Holder, Clear
    100492-862
    Log in for availability
    Each - $25.79
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder for Size 00
    Capsule Holder for Size 00
    100492-864
    Log in for availability
    Each - $31.04
Product Information
  • Catalog #
    Availability
    Price
  • Capsule Holder Multi Task BEEM
    Capsule Holder Multi Task BEEM
    101454-072
    Log in for availability
    Each - $235.41
Product Information
  • Catalog #
    Availability
    Price
  • Cap Holder Set Multi Task BEEM
    Cap Holder Set Multi Task BEEM
    101454-074
    Log in for availability
    Each - $232.13
Product Information
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100488-950
    Log in for availability
    Pack of 4 - $78.00
Product Information
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100488-952
    Log in for availability
    Pack of 6 - $105.72
Product Information
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100488-954
    Log in for availability
    Pack of 12 - $179.46
Product Information
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100492-884
    Log in for availability
    Each - $22.56