Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Paraffin Tissue Microarrays, Electron Microscopy Sciences

Paraffin Tissue Microarrays, Electron Microscopy Sciences

Arraymold Kit C, 15 Cores, 10 Dermal Needles (4 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (4 mm), 1 DVD
Catalog # 102096-522
Supplier #:  69130-40
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Catalog # 102096-522
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Paraffin Tissue Microarrays, Electron Microscopy Sciences

Arraymold Kit C, 15 Cores, 10 Dermal Needles (4 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (4 mm), 1 DVD

Price
$1,648.24
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Paraffin tissue microarrays (TMAs) are becoming more common in the laboratories. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process. Arraymold features include:

  • Allow pathologist or researcher to view many different samples on the same single slide
  • Helpful in finding IHC markers for control or quality assurance from hundreds of samples
  • Staining markers will be more consistent when hundreds of samples are on one slide
  • Used less costly antibody
  • Small enough to be stored in a drawer
  • Rubber arraymold can be reused many times without losing its flexibility

Current techniques including organizing, punching, and inserting tissues is time consuming and involving costly equipments.This Arraymold is the simple tool but great answer for this current technique for laboratories to perform TMAs with less time consuming, excellent results. With this Arraymold, you can process up to 150 specimens onto one single slide with very little time consuming, minimal technical training.

Specifications
  • Description:
    Arraymold Kit C, 15 Cores
  • Includes:
    10 Dermal Needles (4 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (4 mm), 1 DVD
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Styler 4.0 mm
    Styler 4.0 mm
    101764-970
    Log in for availability
    Each - $64.50

Product Information

  • Catalog #
    Availability
    Price
  • Styler 3.5 mm
    Styler 3.5 mm
    101764-968
    Log in for availability
    Each - $64.50

Product Information

  • Catalog #
    Availability
    Price
  • Styler 3.0 mm
    Styler 3.0 mm
    101764-966
    Log in for availability
    Each - $64.50

Product Information

  • Catalog #
    Availability
    Price
  • Styler 1.5 mm
    Styler 1.5 mm
    101764-962
    Log in for availability
    Each - $64.50

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.