Glacier Ice Buckets, Electron Microscopy Sciences
Product 10146-192 is similar to 102096-374 but may have a different manufacturer and/or packaging.
Product Details & Documents
Excellent Insulation: Made of the same triple density expanded polystyrene as our popular Ice Pans, preserve ice 30% longer.
- Won't Sweat - Thick insulating walls will not sweat
- Easy Grip - Robust handles make buckets easy to carry
- Includes Lid - Use the supplied lid to maximize cooling
- Heavy Duty - Solidly constructed to be unbreakable under normal laboratory conditions
- Chemically Resistant - Resistant to mild chemicals* as well as ice-alcohol slurry, dry ice and liquid nitrogen
(*Not recommended for long-term exposure to harsh chemicals or corrosives.)
Specifications
- Top diameter:10.625"