Orders LinkContactUsLinkComponent
Lab-Sorb, Electron Microscopy Sciences
Variety kit Lab-sorb plus variety size bags
  102096-340
 :  68552-10
Lab-Sorb, Electron Microscopy Sciences
  102096-340
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences
Variety kit Lab-sorb plus variety size bags
$92.02
Each
Converts Liquid Waste to Solid Waste for Easy Disposal
Convenient - Includes extra durable disposal bags, super absorbant LabSorb granules and a convenient graduated scoop.
Prevents Spills - Reduces the possibility of accidentally spilling liquid waste in the lab. If spills do occur, use LabSorb for safe liquid containment during cleanup.
Quick - Convert liquids to solids in seconds.
Adaptable - Apply to any volume of liquid waste including 500 mL, 600 mL, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.
Versatile - LabSorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
  • Description:
    Variety kit Lab-sorb plus variety size bags