Orders LinkContactUsLinkComponent
ProScope HR Microscopes, Electron Microscopy Sciences
ProScope HR2 QC Lab
  102096-258
ProScope HR Microscopes, Electron Microscopy Sciences
  102096-258
 :  
ProScope HR Microscopes, Electron Microscopy Sciences
ProScope HR Microscopes, Electron Microscopy Sciences
ProScope HR2 QC Lab
The ProScope HR has been a mainstay in science education, law enforcement, and manufacturing quality control worldwide. The software for Windows and Mac will capture stills, video and time lapse. Lenses range 0 to 10×, 30×, 50×, 100×, 200×, 400× and 1000×. The ProScope HR can be connected to analog microscope, telescope or borescope.
  • Accurate, repeatable measurement within 10 µm
  • Modular lenses range from 1× to 1000×
  • No driver needed, just plug and play
  • Lead-free
  • Attaches to other optics like Borescopes and analog microscopes
  • 30-day no questions asked return policy
  • Two year warranty
The ProScope HR is USB Video Class and the ProScope HR can be used with programs like Skype or iChat to show others what this ProScope HR is seeing in real time over the internet.
The new ProScope HR2 features the new state-of-the-art, Aptina imager which was originally made for surveillance, with large pixels that take in more light than other imagers on the market. The Bodelin engineering team customized this imager to allow extensive imaging adjustment and LED intensity control. The most impressive feature is the ability to actually stream true pixels at higher resolutions.
The ProScope HR2 is the only digital microscope in the world that can do this. Other imagers stream at 640×480 and only interpolate to simulate higher resolutions. The total effect in the ProScope HR2 is live video that appears to be 4 to 5 megapixels in clarity while only using an actual 2 megapixel imager. The new cobalt black handle says this is a serious tool yet the price point is equal to the previous ProScope HR, which will remain in the product line at a lower cost. The ProScope products are currently used in tens of thousands of schools, universities, law enforcement forensic labs, manufacturing quality control, medical and tele-medicine.
Certifications: RoHS compliant.
Delivery information: Software for Windows and Mac is included.
  • Description:
    ProScope HR2 QC Lab
Accessories
  •  
    Description
  • 102096-298
    LX-Proscope HR
    Each - $307.98
  •  
    Description
  • 102096-294
    Screen Measurement Pack Mac Edition
    Each - $148.62
  •  
    Description
  • 102096-292
    Screen Calipers for Windows ProScope Edition
    Each - $58.92
  •  
    Description
  • 102096-284
    ProScope Deluxe Case
    Each - $152.65
  •  
    Description
  • 102096-282
    ProScope Case
    Each - $99.99
  •  
    Description
  • 102096-272
    ProScope C-Mount Adapter
    Each - $46.38
  •  
    Description
  • 102096-216
    ProScope HR2 Base Unit without Lens
    Each - $465.12
  •  
    Description
  • 102096-214
    ProScope HR Base Unit without Lens
    Each - $465.12
  •  
    Description
  • 102096-278
    ProScope Stand
    Each - $307.88
  •  
    Description
  • 102096-290
    MacCalipers ProScope Edition
    Each - $148.62
  •  
    Description
  • 102096-288
    ProScope Image Analyzer Software
    Each - $249.82