PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
PS4R with UKAS Certificate
Catalog # 102096-142
Supplier: Electron Microscopy Sciences
: 68047-PS4R-NA
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
Catalog # 102096-142
Supplier: Electron Microscopy Sciences
CAS Number:
Jump to:
Product Details & Documents
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions.
Specifications
Specifications
- Description:PS4R with UKAS Certificate
- Cat. No.:102096-142