Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Glass Rods, Electron Microscopy Sciences

Glass Rods, Electron Microscopy Sciences

Glass Rods, 200 mm
Catalog # 102095-962
Supplier #:  66206-20
Glass Rods, Electron Microscopy Sciences
Catalog # 102095-962
Glass Rods, Electron Microscopy Sciences
Glass Rods, Electron Microscopy Sciences

Glass Rods, 200 mm

Price
$14.30
Log in for availability
Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Useful tool for all laboratory stirring and mixing

  • 200 mm and 250 mm lengths available
  • Policeman is rubber cover helps help stir at the bottom of container

Useful tool for all laboratories needs to stir/mix substances

Policeman is rubber cover to fit 5 mm rods above. The sealed end is cut at an angle to help stir at the bottom of container.

Glass rods are 5 mm diameter, and come in three lengths.

Specifications
  • Description:
    Glass Rods
  • Length:
    200 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Rubber Policeman
    Rubber Policeman
    102095-968
    Log in for availability
    Pack of 12 - $36.11

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.