Orders LinkContactUsLinkComponent
Single and Multi Wafer Containers, Electron Microscopy Sciences
Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Container Multi, —, 50 mm (2")
Catalog # 102095-800
Supplier #:  64918-2
Single and Multi Wafer Containers, Electron Microscopy Sciences
Catalog # 102095-800
Single and Multi Wafer Containers, Electron Microscopy Sciences
Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Container Multi, —, 50 mm (2")
Price
$118.81
Log in for availability
Pack of 3
Minimum order quantity is 1
Product Details & Documents
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
  • Conical base to support the wafers on their edges
  • Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
Delivery information: The set includes the container, the cover and the spring.
Documents
Specifications
  • Description:
    Wafer Container Multi
  • Size:
  • For use with:
    50 mm (2")