Orders LinkContactUsLinkComponent
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, 18 × 114 mm, 15 ml, 240 mm
Catalog # 102095-648
Supplier #:  64796-15
Tapered Tissue Grinder, Electron Microscopy Sciences
Catalog # 102095-648
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, 18 × 114 mm, 15 ml, 240 mm
Price
$304.37
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require.
  • Homogenizes connective tissue including heart, muscle and lung
  • Made from Wheaton 33 low extractable borosilicate glass
  • Interchangeable components takes the hassle out of matching parts
  • Autoclavable
The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step.The components are interchangeable. Therefore no need keep parts matched during cleaning. The ground glass surface on both the mortar and pestle are capable of homogenizing connective tissue such as heart, muscle and lung, as well as skin and plant tissue.
Specifications
  • Description:
    Tapered Tissue Grinder, 18 × 114 mm
  • Size:
    15 ml
  • Overall length:
    240 mm
Accessories
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Pestle only Tapered Tissue Grinder
    Pestle only Tapered Tissue Grinder
    102095-662
    5 ml
    Log in for availability
    Pack of 2 - $228.56
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Pestle only Tapered Tissue Grinder
    Pestle only Tapered Tissue Grinder
    102095-664
    15 ml
    Log in for availability
    Pack of 2 - $263.38
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Pestle only Tapered Tissue Grinder
    Pestle only Tapered Tissue Grinder
    102095-660
    3 ml
    Log in for availability
    Pack of 2 - $232.94
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Pestle only Tapered Tissue Grinder
    Pestle only Tapered Tissue Grinder
    102095-658
    1 ml
    Log in for availability
    Pack of 2 - $232.94
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Mortar only Tapered Tissue Grinder
    Mortar only Tapered Tissue Grinder
    102095-656
    15 ml
    Log in for availability
    Pack of 2 - $332.45
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Mortar only Tapered Tissue Grinder
    Mortar only Tapered Tissue Grinder
    102095-654
    5 ml
    Log in for availability
    Pack of 2 - $279.71
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Mortar only Tapered Tissue Grinder
    Mortar only Tapered Tissue Grinder
    102095-652
    3 ml
    Log in for availability
    Pack of 2 - $287.76
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Mortar only Tapered Tissue Grinder
    Mortar only Tapered Tissue Grinder
    102095-650
    1 ml
    Log in for availability
    Pack of 2 - $287.76