Orders LinkContactUsLinkComponent
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, 11 × 86 mm, 3 ml, 220 mm
  102095-644
 :  64796-03
Tapered Tissue Grinder, Electron Microscopy Sciences
  102095-644
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, Electron Microscopy Sciences
Tapered Tissue Grinder, 11 × 86 mm, 3 ml, 220 mm
$273.18
Each
Grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require.
  • Homogenizes connective tissue including heart, muscle and lung
  • Made from Wheaton 33 low extractable borosilicate glass
  • Interchangeable components takes the hassle out of matching parts
  • Autoclavable
The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step.The components are interchangeable. Therefore no need keep parts matched during cleaning. The ground glass surface on both the mortar and pestle are capable of homogenizing connective tissue such as heart, muscle and lung, as well as skin and plant tissue.
  • Description:
    Tapered Tissue Grinder, 11 × 86 mm
  • Size:
    3 ml
  • Overall length:
    220 mm
Accessories
  •  
    Description
    Size
  • 102095-662
    Pestle only Tapered Tissue Grinder
    5 ml
    Pack of 2 - $228.56
  •  
    Description
    Size
  • 102095-664
    Pestle only Tapered Tissue Grinder
    15 ml
    Pack of 2 - $263.38
  •  
    Description
    Size
  • 102095-660
    Pestle only Tapered Tissue Grinder
    3 ml
    Pack of 2 - $232.94
  •  
    Description
    Size
  • 102095-658
    Pestle only Tapered Tissue Grinder
    1 ml
    Pack of 2 - $232.94
  •  
    Description
    Size
  • 102095-656
    Mortar only Tapered Tissue Grinder
    15 ml
    Pack of 2 - $332.45
  •  
    Description
    Size
  • 102095-654
    Mortar only Tapered Tissue Grinder
    5 ml
    Pack of 2 - $279.71
  •  
    Description
    Size
  • 102095-652
    Mortar only Tapered Tissue Grinder
    3 ml
    Pack of 2 - $287.76
  •  
    Description
    Size
  • 102095-650
    Mortar only Tapered Tissue Grinder
    1 ml
    Pack of 2 - $287.76