Orders LinkContactUsLinkComponent
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Tissue Grinder Pestle only PTFE Radial Serrations, 15 ml
Catalog # 102095-636
 :  64795-83
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Catalog # 102095-636
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Tissue Grinder Pestle only PTFE Radial Serrations, 15 ml
Price
$321.90
Log in for availability
Pack of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All features are similar to the Potter-Elvehjem Tissue Grinder series, with additional radial serrated PTFE pestle.
This grinder has a serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm (0.20 to 0.25”).
Specifications
  • Description:
    Tissue Grinder Pestle only PTFE Radial Serrations
  • Size:
    15 ml