Orders LinkContactUsLinkComponent
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Tissue Grinder Pestle only PTFE Radial Serrations, 2 ml
  102095-630
 :  64795-80
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
  102095-630
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Tissue Grinder Pestle only PTFE Radial Serrations, 2 ml
$284.51
Pack of 2
All features are similar to the Potter-Elvehjem Tissue Grinder series, with additional radial serrated PTFE pestle.
This grinder has a serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm (0.20 to 0.25”).
  • Description:
    Tissue Grinder Pestle only PTFE Radial Serrations
  • Size:
    2 ml