Orders LinkContactUsLinkComponent
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Potter-Elvehjem Micro Tissue Grinder, 0.1 ml
  102095-612
 :  64794-01
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
  102095-612
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Potter-Elvehjem Micro Tissue Grinder, 0.1 ml
$287.46
Pack of 2
Similar to the all glass Potter-Elvehjem style, but micro size. Ideal for extremely precise work.
  • Made from Wheaton 33 low extractable borosilicate glass
  • Micro size for extremely precise work
  • Reservoir and pouring lip are incorporated into the design
  • Autoclavable
This unit is designed for delicate hand operation. Clearance between pestle and tube is 0.004” to 0.006” (0.1 – 015 mm).
  • Description:
    Potter-Elvehjem Micro Tissue Grinder
  • Size:
    0.1 ml