Orders LinkContactUsLinkComponent
Tri-Seal™ container, Electron Microscopy Sciences
Tri-Seal™ container, Electron Microscopy Sciences
50 ml, Tall With Locking Latch, 28 × 84 mm
Catalog # 102095-000
Supplier #:  64235-50TL
Tri-Seal™ container, Electron Microscopy Sciences
Catalog # 102095-000
Tri-Seal™ container, Electron Microscopy Sciences
Tri-Seal™ container, Electron Microscopy Sciences
50 ml, Tall With Locking Latch, 28 × 84 mm
Price
$1,712.16
Log in for availability
Pack of 600
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
Specifications
  • Capacity:
    50 ml
  • Description:
    Tall With Locking Latch
  • Dimensions:
    28 × 84 mm
  • Cat. No.:
    102095-000