FlexTainer Containers, Electron Microscopy Sciences
Product Details & Documents
This is a space-saving expandable accordion-style jug with flattens down for easy transport and storage.
Ideal for storing and carrying numerous types of fluids, it is suitable for any laboratory. Made of high strength polyethylene, it features comfortable carry grip and screw-on cap with on/off spout. It is available in many sizes up to 8 liters. Come with individually wrapped.
Specifications
- Capacity:8 L
- Description:Flex-Tainer™ Accordion-style Jug