Orders LinkContactUsLinkComponent
ProPlate™ MicroArray System, Electron Microscopy Sciences
4 Well Module, 16x16 mm Well
  102094-660
 :  63484-24
ProPlate™ MicroArray System, Electron Microscopy Sciences
  102094-660
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
4 Well Module, 16x16 mm Well
$383.42
Pack of 10
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
  • Description:
    4 Well Module, 16x16 mm Well