ProPlate™ MicroArray System, Electron Microscopy Sciences
Product Details & Documents
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
Specifications
- Description:2 Well ProPlate Module Tray Set, 4-16.5x34 mm Well, Delrin Snap Clips