Orders LinkContactUsLinkComponent
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
16 Well ProPlate Slide Module Tray Set, 3-6 mm dia, Stainless Steel Spring Clips
  102094-632
Supplier #:  63484-06
ProPlate™ MicroArray System, Electron Microscopy Sciences
  102094-632
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
16 Well ProPlate Slide Module Tray Set, 3-6 mm dia, Stainless Steel Spring Clips
$271.40
Each
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
  • Description:
    16 Well ProPlate Slide Module Tray Set, 3-6 mm dia, Stainless Steel Spring Clips