Orders LinkContactUsLinkComponent
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Slide Drying Rack, Side mounted, —
Catalog # 102094-544
 :  63214-55
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Catalog # 102094-544
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Slide Drying Rack, Side mounted, —
Price
$173.51
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This Flotation Workstation helps to make your tissue preparation easier, safer and more efficient.
  • Magnetic non-contact switch function for years flawlessly
  • Microprocessor-controlled heating, ambient to 65°C
  • Flat membrane switches for safety and simplicity
  • Fluorescent white lighting positioned for improve tissue viewing glare-free
  • Heat-resistant plastic housing maintains cosmetic appearance
  • Removable glass dish for easy cleaning
  • 2 x 6" or 2 x 8" Convection Slide dryer & 2 x 2" HISTO Orientator™ for eliminating wrinkles
The unit comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled electronics and a high contrast background for optimum specimen viewing combined with the HISTO Orientator™ for flattening sections and a Slide Dryer, all in one compact unit. The systems are now available in Low Profile 8 x 8 x 2 ¼" deep dish versions as well as for those labs wanting to float larger ribbons, 8 x 11 x 2".
For those flotation bath applications not requiring the add convenience of Slide Dryer and HISTO Orientator™, the basic Flotation Bath provides all the functional need of the Flotation Workstation at the affordable price. The baths again come in two sizes , 8 x 8 x 2 1/4" or 8 x 11 x 2".
Specifications
  • Description:
    Slide Drying Rack, Side mounted
  • Size:
Some Products May Appear Restricted