Orders LinkContactUsLinkComponent
Strip Disposable Mold, Electron Microscopy Sciences
Strip Disposable Mold, Electron Microscopy Sciences
37 x 24 x 7 mm
Catalog # 102094-028
Strip Disposable Mold, Electron Microscopy Sciences
Catalog # 102094-028
Supplier:  
Strip Disposable Mold, Electron Microscopy Sciences
Strip Disposable Mold, Electron Microscopy Sciences
37 x 24 x 7 mm
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BASE MOLD STRIP DISP PS 30X24X5MM PK100 Catalog #102094-024
Product 102094-024 is a functional alternative to 102094-028 but Packaging, Quantity, Manufacturer and/or specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Strip disposable molds are manufactured from high impact polystyrene, offering a low cost alternative to stainless steel
These molds are available in a choice of sizes and depths. The molds have a smooth interior finish and rounded corners for ease of removal. There are five molds per strip and they are packed 100 strips to a case.
Specifications
  • Size:
    37 x 24 x 7 mm