Strip Disposable Mold, Electron Microscopy Sciences
Product Details & Documents
Strip disposable molds are manufactured from high impact polystyrene, offering a low cost alternative to stainless steel
These molds are available in a choice of sizes and depths. The molds have a smooth interior finish and rounded corners for ease of removal. There are five molds per strip and they are packed 100 strips to a case.
Specifications
- Size:30 x 24 x 5 mm