Orders LinkContactUsLinkComponent
True North™ Cool Containers, Electron Microscopy Sciences
True North™ Cool Containers, Electron Microscopy Sciences
Cool Containers, 4L Bucket, Black Onyx
Catalog # 102093-894
Supplier #:  62013-04
True North™ Cool Containers, Electron Microscopy Sciences
Catalog # 102093-894
True North™ Cool Containers, Electron Microscopy Sciences
True North™ Cool Containers, Electron Microscopy Sciences
Cool Containers, 4L Bucket, Black Onyx
Price
$179.27
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These rigid single mold polyurethane containers provides durability for chilling your temperature sensitive samples. Excellent insulation properties for ultra colduse with ice, LN2, ice-salt slurries, and more.
  • Convenient lid nests beneath bucket or pan and easily removes with comfortable recessed knobs
  • Snug fit minimizes evaporation loss
  • Handy pour spout for pouring melted ice, water, and solutions
  • Ergonomic handles safely transport bucket or pan from work area to sink
  • Deep internal cavity has enough room to add a rack
  • Tapered sides make stacking and storage compact
  • Available in a bucket or pan and in multiple sizes
Specifications
  • Description:
    Cool Containers, 4L Bucket, Black Onyx