Orders LinkContactUsLinkComponent
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Natural
Catalog # 102093-624
Supplier #:  61600-N
81-Well Storage Rack, Electron Microscopy Sciences
Catalog # 102093-624
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Natural
Price
$100.23
Log in for availability
Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Durable polypropylene rack fits in standard 5.25” square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials.
  • Molded grid lines on the lid
  • Imprinted coordinates on the bottom of base
  • Molded reference point on the lid
  • Rack measures 5.125×5.125×1.75 cm
Specifications
  • Color:
    Natural
  • Description:
    81-Well Storage Rack