Orders LinkContactUsLinkComponent
Calibration Micrometer, Electron Microscopy Sciences
Calibration Micrometer, Electron Microscopy Sciences
Model ACM-1, Calibrating Micrometer
Catalog # 102093-154
Supplier #:  60218-1PG
Calibration Micrometer, Electron Microscopy Sciences
Catalog # 102093-154
Calibration Micrometer, Electron Microscopy Sciences
Calibration Micrometer, Electron Microscopy Sciences
Model ACM-1, Calibrating Micrometer
Price
$661.13
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This ideal standard provides accurate calibration of any eyepiece, reticules, filar micrometers, or electronic measuring instrument
Features a series of ʽH’ shaped fiducial images in a range of sizes from 0.1 mm through 20 mm, useful for calibration range from 1X to 1000X power.
Specifications
  • Description:
    Model ACM-1, Calibrating Micrometer