Microscope Cameras, Electron Microscopy Sciences
Product Details & Documents
We offer three types of camera systems for documenting resolved microscopic images.
All components are conveniently packaged under one part number for easy ordering.
Specifications
- Resolution:3 MP
- Sensor type:CMOS
- Description:Digital Video Color Camera USB 2.0, for upright compound microscopes