Orders LinkContactUsLinkComponent
Neat Stain™ Stain Kits, Electron Microscopy Sciences
Neat Stain™ Stain Kits, Electron Microscopy Sciences
Trichrome Stain Kit
Catalog # 102092-394
Neat Stain™ Stain Kits, Electron Microscopy Sciences
Catalog # 102092-394
Supplier:  
CAS Number:  
Neat Stain™ Stain Kits, Electron Microscopy Sciences
Neat Stain™ Stain Kits, Electron Microscopy Sciences
Trichrome Stain Kit
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Neat Stain™ Kits are specifically designed for stat procedures and/or laboratories using manual staining methods. Neat Stain™ is available as a rapid three-step differential hematology stain, Gram stain and Trichrome stain staining kit. They are quite simple to use, just immerse your slides sequentially in each reagent for the correct time, air dry and read.
  • Neat – Pre-filled multi-well vessels with self-contained foil sealed
  • Fast – Immerse slides sequentially in each reagent for specified time, air dry and read
  • Convenient – Additional foils for re-sealing containers
Neat Stain™ GRAM Kit is a 4-step stain kit is prepared for staining bacteria from cultures or specimens by the differential Gram stain method. This kit stains similar to the traditional Wright's and Wright-Giemsa stains. Neat Stain™ offers the flexibility of the three dip staining sequence in both the stain and counter stain solution.
Neat Stain™ Hematology Stain Kit is 3-step stain procedure is designed for differentiation of morphological cell types in peripheral blood smears. This kit is also useful in andrology labs staining microorganism and spermatozoa. Staining characteristics are similar to the Wrights and Wright-Giemsa stains. Neat Stain offers the flexibility of a three dip staining format in both the stain and counter stain solution.
Certifications: Certificates of analysis are available by lot or product number.
Ordering information: Slide Holder sold separately.
Delivery information: All kits have one plastic stand and 6 re-seal foils. GRAM and Hematology Kits have 6 Reagent Packs and performs 150 tests. Trichrome kit has 3 reagent packs and performs 75 tests.
Specifications
Specifications
  • Description:
    Trichrome Stain Kit