Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Michel's Buffer, Electron Microscopy Sciences

Michel's Buffer, Electron Microscopy Sciences

Michel's Buffer, 500 ml, Bottle
Catalog # 102092-164
Supplier #:  18041-05
Michel's Buffer, Electron Microscopy Sciences
Catalog # 102092-164
CAS Number:  
Michel's Buffer, Electron Microscopy Sciences
Michel's Buffer, Electron Microscopy Sciences

Michel's Buffer, 500 ml, Bottle

CAS Number:  
Price
$49.29
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Michel’s Buffer is available ready-to-use for your convenience.

  • Store at room temperature
Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Michel's Buffer
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.