Orders LinkContactUsLinkComponent
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
Lower GI, 12 Compartments
Catalog # 102091-900
Supplier #:  15736-40
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
Catalog # 102091-900
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
Lower GI, 12 Compartments
Price
$124.46
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
We are proud to offer a multi-compartment container in the shape of a dish and half prefilled with 10% Neutral buffered Formalin, for holding and transporting biopsies
It is supplied with a leak-proof closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. These CoreDisn™ measure only 15 x 95 mm. Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted. Thanks to CoreDish™ it is no more necessary to use a multiple of individual containers, thereby reducing cost and risks of confusion. Few recommendations concerning how the biopsies should be handled have been published. Performing a large number of biopsies means an increase in the number of containers handled and consequently a technical overload of the transmission network, which occurs without any financial counterpart. A new approach had to be developed in order to increase productivity.
We are proud to offer a multi-compartment container in the shape of a dish and half prefilled with 10% Neutral buffered Formalin, for holding and transporting biopsies. It is supplied with a leak-proof closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. These CoreDisn™ measure only 15 x 95 mm. Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted. Thanks to CoreDish™ it is no more necessary to use a multiple of individual containers, thereby reducing cost and risks of confusion.
CoreDish™ available in many configurations to serve in different biopsy, such as prostate, upper GI tract and lower GI tract. A label allows recording the related information, patient ID, doctor, date and time.
M970-D5B-2 CoreDish™ Breast Biopsy Container is half prefilled with 10% Formalin. This multi-compartment container (out of five compartments, four are labeled: LUQ, RUQ, LLQ, RLQ) in the shape of a dish and half prefilled with 10% Neutral Buffered Formalin, for holding and transporting biopsies. It is supplied with a leak-proof screw closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. The dish measures only 15 x 95 mm in diameter. Each compartment is clearly identified to allow proper placement and visualization of the breast biopsy being inserted. A writing area for patient information is provided.
M970-D8P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled com partments: Base, Lateral Base, Medium, Lateral Medium, Apex, Lateral Apex.
M970-D8UGI CoreDish™ Upper GI Biopsy Container is half prefilled with 10% Formalin. For upper GI track biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled compartments: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Esophage, Pylorus, Duodeum.
M970-D8LGI CoreDish™ Lower Gi Biopsy Container is half prefilled with 10% Formalin. For lower GI track biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Ten labeled compartments: Proximal Flexure Colon, Hepatic Fleure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal lleum, Rectum, Sigmmoid Colon.
M970-D12P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. TWelve labeled compartments: L Base, R Base, L Lateral Base, R Lateral Base, L Medium, R Medium, L Lateral Medium, R Lateral Medium, L Lateral , L Apex, R Apex, R Lateral Apex. CoreDish™ available in many configurations to serve in different biopsy, such as prostate, upper GI tract and lower GI tract. A label allows recording the related information, patient ID, doctor, date and time.
M970-D5B-2 CoreDish™ Breast Biopsy Container is half prefilled with 10% Formalin. This multi-compartment container (out of five compartments, four are labeled: LUQ, RUQ, LLQ, RLQ) in the shape of a dish and half prefilled with 10% Neutral Buffered Formalin, for holding and transporting biopsies. It is supplied with a leak-proof screw closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. The dish measures only 15 x 95 mm in diameter. Each compartment is clearly identified to allow proper placement and visualization of the breast biopsy being inserted. A writing area for patient information is provided.
M970-D8P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled com partments: Base, Lateral Base, Medium, Lateral Medium, Apex, Lateral Apex.
M970-D8UGI CoreDish™ Upper GI Biopsy Container is half prefilled with 10% Formalin. For upper GI track biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled compartments: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Esophage, Pylorus, Duodeum.
M970-D8LGI CoreDish™ Lower Gi Biopsy Container is half prefilled with 10% Formalin. For lower GI track biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Ten labeled compartments: Proximal Flexure Colon, Hepatic Fleure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal lleum, Rectum, Sigmmoid Colon.
M970-D12P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. TWelve labeled compartments: L Base, R Base, L Lateral Base, R Lateral Base, L Medium, R Medium, L Lateral Medium, R Lateral Medium, L Lateral , L Apex, R Apex, R Lateral Apex.
Specifications
  • Description:
    Lower GI, 12 Compartments