Orders LinkContactUsLinkComponent
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Paraformaldehyde Solution and glut, 1 L
Catalog # 102091-842
Supplier #:  15732-10
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Catalog # 102091-842
CAS Number:  
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Paraformaldehyde Solution and glut, 1 L
CAS Number:  
Price
$617.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Fixative Reagent Grade (Karnovsky’s). This fixative contains 3% glutaraldehyde and 2% formaldehyde in 0.1M phosphate buffer, pH 7.4. Karnovsky’s with formalin 2% and Biological glutaraldehyde 3% in 0.1m Phosphate Buffer. Karnovsky’s with paraformaldehyde solution and EM grade glutaraldehyde.
Specifications
Specifications
  • Description:
    Karnovsky Fixative Paraformaldehyde Solution and glut
  • Size:
    1 L