INFINITY 2-1 C CCD Color Camera, Electron Microscopy Sciences
Product Details & Documents
Equipped with a high-quality, Sony CCD sensor, INFINITY CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range and a 12-bit digital output.
- 1, 2 and 3 megapixel resolution
- Higher dynamic range for quantitative analysis
- Fast frame rates
- Low noise electronics
Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest. It is ideal for higherend scientific, medical, ophthalmic and life science applications.
Applications: Brightfield, Darkfield, DIC, Live Cell Imaging, Histology, Pathology, Cytology, Defect Analysis, Semiconductor Inspection, Metrology, Gel Documentation, and Moderate Light Fluorescence.
Specifications
- Resolution:3.3 MP
- Sensor type:CCD
- Description:INFINITY2-3C Color Camera
- Interfaces:USB 2.0
- Cat. No.:102091-458
- Sensor size:1/1.8"
- Model:INFINITY2-3C
- Frame Rate:5 fps
- Sensor:CCD Color